siRNA Details
| Gene Symbol: Nln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029447.2 | NP_083723.1 | 3 | 549 | ||
| GenBank mRNA | AK079090.1 | - | 3 | 550 | ||
| AK138399.1 | - | 3 | 522 | |||
| AK146364.1 | - | 3 | 498 | |||
| AK170508.1 | - | 3 | 522 | |||
| AK171432.1 | - | 3 | 522 | |||
| BC016224.1 | - | 3 | 522 | 4486941 | ||

Nln
Genomic Map