siRNA Details
| Gene Symbol: Smc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133786.3 | NP_598547.1 | 6 | 871 | ||
| GenBank mRNA | AJ534940.2 | - | 5 | 747 | ||
| AK010848.1 | - | 4 | 322 | |||
| AK048650.1 | - | 1 | 83 | |||
| AK088350.1 | - | 6 | 854 | |||
| AK088846.1 | - | 6 | 837 | |||
| AK133817.1 | - | 6 | 861 | |||
| AK152804.1 | - | 6 | 898 | |||
| BC061481.1 | - | 6 | 846 | 6512832 | ||
| BC062939.1 | - | 6 | 871 | 6841276 | ||

Smc4
Genomic Map