siRNA Details
| Gene Symbol: Farp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134082.3 | NP_598843.3 | 26 | 3421 | ||
| GenBank mRNA | AK034903.1 | - | 7 | 724 | ||
| BC004009.1 | - | 10 | 1162 | |||
| BC027077.1 | - | 9 | 964 | |||
| BC030329.1 | - | 19 | 2409 | |||
| BC043327.1 | - | 8 | 825 | |||
| BC141229.1 | - | 25 | 2982 | |||

Farp1
Genomic Map