siRNA Details
| Gene Symbol: Samd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146025.2 | NP_666137.1 | 7 | 1394 | ||
| GenBank mRNA | AK044765.1 | - | 7 | 923 | ||
| AK047184.1 | - | 7 | 1371 | |||
| AK049753.1 | - | 7 | 1397 | |||
| BC034054.1 | - | 7 | 918 | 4505400 | ||
| BC049954.1 | - | 7 | 951 | 4191611 | ||

Samd14
Genomic Map