siRNA Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170673.3 | NP_733773.2 | 21 | 1720 | ||
| GenBank mRNA | AK042692.1 | - | 21 | 1721 | ||
| AK044080.1 | - | 21 | 1686 | |||
| AK047571.1 | - | 19 | 2735 | |||
| AK137422.1 | - | 21 | 1721 | |||
| AK220550.1 | - | 13 | 3084 | |||
| BC115728.1 | - | 21 | 1670 | 40039295 | ||
| BQ550678.1 | - | 5 | 355 | |||

Cpne9
Genomic Map