siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175031.3 | NP_778196.2 | 4 | 509 | ||
| GenBank mRNA | AK007188.1 | - | 4 | 452 | ||
| AK029842.1 | - | 4 | 485 | |||
| AK173142.1 | - | 4 | 469 | |||
| BC043103.1 | - | 4 | 461 | 6406485 | ||
| BC049721.1 | - | 4 | 435 | |||
| BC058698.1 | - | 4 | 574 | 6831702 | ||
| BC061470.1 | - | 3 | 318 | |||
| CK633987.1 | - | 4 | 513 | |||

Stk36
Genomic Map