siRNA Details
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175090.4 | NP_780299.2 | 4 | 462 | ||
GenBank mRNA | AK046835.1 | - | 4 | 415 | ||
AK151337.1 | - | 4 | 444 | |||
AK170285.1 | - | 4 | 415 | |||
BC034674.1 | - | 4 | 400 | 3968084 | ||
BC046470.1 | - | 2 | 136 | |||
BC058227.1 | - | 4 | 402 | 6309323 | ||
BC065147.1 | - | 3 | 310 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133600, BC078745, AF268030, AY539951.
Panther Classification:
Gene Ontology Categories:
