siRNA Details
| Gene Symbol: Rnf122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175136.2 | NP_780345.1 | 2 | 443 | ||
| GenBank mRNA | AK077856.1 | - | 2 | 463 | ||
| AK085376.1 | - | 2 | 425 | |||
| AK136206.1 | - | 2 | 461 | |||
| AK162034.1 | - | 2 | 461 | |||
| AK171823.1 | - | 2 | 463 | |||
| BC038399.1 | - | 2 | 443 | 4947596 | ||
| BC050268.1 | - | 1 | 130 | |||

Rnf122
Genomic Map