siRNA Details
| Gene Symbol: Cyp4a12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177406.3 | NP_803125.2 | 6 | 750 | ||
| GenBank mRNA | AK144260.1 | - | 6 | 680 | ||
| BC014721.1 | - | 6 | 696 | 4240359 | ||
| BC025936.1 | - | 6 | 690 | 5097698 | ||
| BC026582.1 | - | 6 | 696 | 4971335 | ||
| BC031141.1 | - | 6 | 671 | 4972684 | ||
| BC033924.1 | - | 6 | 680 | 5101821 | ||

Cyp4a12a
Genomic Map