siRNA Details
| Gene Symbol: Aqp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159658.1 | NP_001153130.1 | 1 | 145 | ||
| NM_177587.2 | NP_808255.1 | 1 | 145 | |||
| GenBank mRNA | AB084104.1 | - | 1 | 208 | ||
| AK131815.1 | - | 1 | 146 | |||
| BC107212.1 | - | 1 | 139 | 40051607 | ||
| BC107213.1 | - | 1 | 137 | 40051613 | ||

Aqp12
Genomic Map