siRNA Details
| Gene Symbol: Zfp281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160251.1 | NP_001153723.1 | 2 | 831 | ||
| NM_177643.4 | NP_808311.1 | 2 | 827 | |||
| GenBank mRNA | AK142218.1 | - | 2 | 744 | ||
| AK157189.1 | - | 2 | 770 | |||
| BC003243.1 | - | 2 | 788 | 3498439 | ||
| BC062153.1 | - | 2 | 781 | 6335572 | ||
| CT010200.1 | - | 1 | 662 | |||

Zfp281
Genomic Map