siRNA Details
| Gene Symbol: Bloc1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177692.3 | NP_808360.2 | 2 | 140 | ||
| GenBank mRNA | AK054006.1 | - | 2 | 159 | ||
| AK153811.1 | - | 177 | ||||
| BC043666.1 | - | 2 | 140 | 5025443 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bloc1s3| Gene Symbol: Bloc1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177692.3 | NP_808360.2 | 2 | 140 | ||
| GenBank mRNA | AK054006.1 | - | 2 | 159 | ||
| AK153811.1 | - | 177 | ||||
| BC043666.1 | - | 2 | 140 | 5025443 | ||