siRNA Details
| Gene Symbol: Mterf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178051.3 | NP_835152.1 | 3 | 588 | ||
| GenBank mRNA | AK007901.1 | - | 3 | 575 | ||
| AK077142.1 | - | 3 | 588 | |||
| AK079098.1 | - | 3 | 580 | |||
| BC002280.1 | - | 2 | 201 | 3497421 | ||
| BC055877.1 | - | 2 | 532 | 4236729 | ||
| BC064709.1 | - | 3 | 581 | 30314353 | ||

Mterf4
Genomic Map