siRNA Details
| Gene Symbol: ING2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291959.1 | NP_001278888.1 | 2 | 652 | ||
| NM_001564.3 | NP_001555.1 | 2 | 733 | |||
| XM_011531927.2 | XP_011530229.1 | 2 | 578 | |||
| GenBank mRNA | AB012853.1 | - | 2 | 678 | ||
| AB196793.1 | - | 2 | 527 | |||
| AF053537.1 | - | 2 | 654 | |||
| AJ006851.1 | - | 2 | 587 | |||
| AK294310.1 | - | 2 | 674 | |||
| AK314000.1 | - | 2 | 734 | |||
| BC030128.2 | - | 2 | 713 | 3941655 | ||

ING2
Genomic Map