siRNA Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001565.3 | NP_001556.2 | 4 | 540 | ||
GenBank mRNA | BC010954.1 | - | 4 | 505 | 4274617 | |
X02530.1 | - | 4 | 501 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001565.3 | NP_001556.2 | 4 | 540 | ||
GenBank mRNA | BC010954.1 | - | 4 | 505 | 4274617 | |
X02530.1 | - | 4 | 501 |