siRNA Details
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174991.3 | NP_778156.2 | 2 | 1317 | ||
GenBank mRNA | AK147456.1 | - | 2 | 1302 | ||
AK147459.1 | - | 2 | 1270 | |||
AK147494.1 | - | 2 | 1104 | |||
AK147600.1 | - | 2 | 1079 | |||
AK147607.1 | - | 2 | 1321 | |||
AK220481.1 | - | 2 | 682 | |||
AY168408.1 | - | 2 | 829 | |||
BC115835.1 | - | 2 | 823 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
