siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175031.3 | NP_778196.2 | 5 | 661 | ||
| GenBank mRNA | AK007188.1 | - | 5 | 604 | ||
| AK029842.1 | - | 5 | 637 | |||
| AK173142.1 | - | 5 | 621 | |||
| BC043103.1 | - | 5 | 613 | 6406485 | ||
| BC049721.1 | - | 5 | 587 | |||
| BC058698.1 | - | 5 | 726 | 6831702 | ||
| BC061470.1 | - | 4 | 470 | |||
| CK633987.1 | - | 5 | 665 | |||

Stk36
Genomic Map