siRNA Details
| Gene Symbol: Tmem161b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175187.5 | NP_780396.2 | 7, 8 | 804 | ||
| GenBank mRNA | AK013305.1 | - | 6, 7 | 633 | ||
| AK088393.1 | - | 7, 8 | 776 | |||
| BC037611.1 | - | 5, 6 | 584 | |||
| BC051663.1 | - | 7, 8 | 752 | 30020101 | ||
| BC060067.1 | - | 7, 8 | 786 | |||

Tmem161b
Genomic Map