siRNA Details
| Gene Symbol: Ccdc88a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176841.4 | NP_789811.2 | 21 | 3853 | ||
| GenBank mRNA | AB087827.1 | - | 21 | 3677 | ||
| AK129310.1 | - | 8 | 1841 | |||
| BC037020.1 | - | 3 | 422 | |||
| BC079895.1 | - | 12 | 2699 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008770470.

Ccdc88a
Genomic Map