siRNA Details
| Gene Symbol: Mink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045959.1 | NP_001039424.1 | 3 | 359 | ||
| NM_001045964.1 | NP_001039429.1 | 3 | 359 | |||
| NM_016713.2 | NP_057922.2 | 3 | 359 | |||
| NM_176893.2 | NP_795712.2 | 3 | 359 | |||
| GenBank mRNA | AB035697.1 | - | 3 | 344 | ||
| AB041925.1 | - | 3 | 345 | |||
| BC052474.1 | - | 3 | 223 | 6833893 | ||
| BC079549.1 | - | 3 | 217 | |||
| CB245214.1 | - | 3 | 223 | |||
| U49949.1 | - | 1 | 80 | |||

Mink1
Genomic Map