siRNA Details
| Gene Symbol: Cnksr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177751.2 | NP_808419.1 | 2 | 467 | ||
| GenBank mRNA | AK122399.1 | - | 2 | 538 | ||
| AK141544.1 | - | 2 | 538 | |||
| BC043093.1 | - | 2 | 467 | 6405389 | ||
| BC060716.1 | - | 2 | 467 | 6405389 | ||

Cnksr2
Genomic Map