siRNA Details
| Gene Symbol: Mterf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178051.3 | NP_835152.1 | 3 | 638 | ||
| GenBank mRNA | AK007901.1 | - | 3 | 625 | ||
| AK077142.1 | - | 3 | 638 | |||
| AK079098.1 | - | 3 | 630 | |||
| BC002280.1 | - | 2 | 251 | 3497421 | ||
| BC055877.1 | - | 2 | 582 | 4236729 | ||
| BC064709.1 | - | 3 | 631 | 30314353 | ||

Mterf4
Genomic Map