siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175031.3 | NP_778196.2 | 14, 15 | 2007 | ||
| GenBank mRNA | AK029842.1 | - | 14, 15 | 1917 | ||
| AK047284.1 | - | 3, 4 | 460 | |||
| AK173142.1 | - | 13, 14 | 1873 | |||
| BC043103.1 | - | 14, 15 | 1959 | 6406485 | ||
| BC061470.1 | - | 13, 14 | 1816 | |||

Stk36
Genomic Map