siRNA Details
| Gene Symbol: Tsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177325.3 | NP_796299.2 | 7 | 1293 | ||
| GenBank mRNA | AK030545.1 | - | 6 | 1199 | ||
| AK129350.1 | - | 7 | 1247 | |||
| AK154063.1 | - | 7 | 1280 | |||
| AK162686.1 | - | 1 | 79 | |||
| BC031531.1 | - | 7 | 1266 | 4014819 | ||

Tsr1
Genomic Map