siRNA Details
| Gene Symbol: Vps37b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177876.4 | NP_808544.1 | 2 | 311 | ||
| GenBank mRNA | AK082658.1 | - | 2 | 272 | ||
| AK166359.1 | - | 2 | 292 | |||
| AK172114.1 | - | 2 | 272 | |||
| AK172337.1 | - | 2 | 272 | |||
| BC026744.1 | - | 2 | 312 | 4163734 | ||

Vps37b
Genomic Map