siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178065.4 | NP_835166.3 | 5 | 997 | ||
| GenBank mRNA | AB093230.1 | - | 4 | 616 | ||
| AK077015.1 | - | 4 | 529 | |||
| AK133332.1 | - | 4 | 531 | |||
| AK147491.1 | - | 5 | 998 | |||
| AK147878.1 | - | 5 | 961 | |||
| AK154529.1 | - | 5 | 978 | |||
| BC057351.1 | - | 5 | 935 | |||
| BC060658.1 | - | 4 | 566 | 30355406 | ||
| BC079590.1 | - | 5 | 955 | |||

Arel1
Genomic Map