siRNA Details
| Gene Symbol: Wdr92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178909.4 | NP_849240.1 | 2 | 271 | ||
| GenBank mRNA | AK033294.1 | - | 2 | 217 | ||
| AK047691.1 | - | 2 | 246 | |||
| AK088218.1 | - | 2 | 246 | |||
| AK142228.1 | - | 2 | 217 | |||
| BC096540.1 | - | 2 | 200 | 6403681 | ||
| BY794211.1 | - | 2 | 272 | |||

Wdr92
Genomic Map