siRNA Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178911.4 | NP_849242.1 | 5 | 578 | ||
| GenBank mRNA | AK045459.1 | - | 5 | 537 | ||
| AK079549.1 | - | 5 | 537 | |||
| AK079878.1 | - | 5 | 580 | |||
| AK151551.1 | - | 5 | 812 | |||
| AK152969.1 | - | 4 | 425 | |||
| AK170362.1 | - | 5 | 535 | |||
| BC058565.1 | - | 5 | 528 | 6442038 | ||

Pld4
Genomic Map