siRNA Details
| Gene Symbol: Aftph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252503.2 | NP_001239432.1 | 2 | 459 | ||
| NM_001290545.1 | NP_001277474.1 | 2 | 459 | |||
| NM_181411.4 | NP_852076.1 | 2 | 459 | |||
| GenBank mRNA | AK029681.1 | - | 2 | 358 | ||
| AK030572.1 | - | 2 | 403 | |||
| AK042681.1 | - | 2 | 397 | |||
| AK138793.1 | - | 2 | 381 | |||
| BC052036.1 | - | 2 | 387 | 5702534 | ||

Aftph
Genomic Map