siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178065.4 | NP_835166.3 | 8 | 1514 | ||
| GenBank mRNA | AB093230.1 | - | 7 | 1133 | ||
| AK077015.1 | - | 7 | 1046 | |||
| AK133332.1 | - | 7 | 1049 | |||
| AK147491.1 | - | 8 | 1515 | |||
| AK147878.1 | - | 8 | 1478 | |||
| AK154529.1 | - | 8 | 1495 | |||
| BC049900.1 | - | 2 | 204 | |||
| BC057351.1 | - | 8 | 1452 | |||
| BC060658.1 | - | 7 | 1083 | 30355406 | ||
| BC079590.1 | - | 8 | 1472 | |||

Arel1
Genomic Map