siRNA Details
| Gene Symbol: Lingo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181074.4 | NP_851419.1 | 4 | 1148 | ||
| GenBank mRNA | AF373780.1 | - | 1 | 700 | ||
| AK027262.2 | - | 2 | 771 | |||
| AK034084.1 | - | 2 | 1248 | |||
| BC052384.1 | - | 4 | 1148 | 5703857 | ||
| BC065696.1 | - | 4 | 1093 | 5685897 | ||

Lingo1
Genomic Map