siRNA Details
Gene Symbol: B230219D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181278.2 | NP_851795.1 | 2 | 602 | ||
GenBank mRNA | AJ492504.1 | - | 1 | 393 | ||
BC051430.1 | - | 2 | 602 | 6530869 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B230219D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181278.2 | NP_851795.1 | 2 | 602 | ||
GenBank mRNA | AJ492504.1 | - | 1 | 393 | ||
BC051430.1 | - | 2 | 602 | 6530869 |