siRNA Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181345.3 | NP_851990.2 | 3 | 284 | ||
| GenBank mRNA | AK054533.1 | - | 3 | 285 | ||
| AK139497.1 | - | 3 | 285 | |||
| AY262112.1 | - | 3 | 284 | |||
| BC048751.1 | - | 3 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npm2| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181345.3 | NP_851990.2 | 3 | 284 | ||
| GenBank mRNA | AK054533.1 | - | 3 | 285 | ||
| AK139497.1 | - | 3 | 285 | |||
| AY262112.1 | - | 3 | 284 | |||
| BC048751.1 | - | 3 | 259 | |||