siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178065.4 | NP_835166.3 | 9 | 1735 | ||
| GenBank mRNA | AB093230.1 | - | 8 | 1354 | ||
| AK077015.1 | - | 8 | 1267 | |||
| AK133332.1 | - | 8 | 1271 | |||
| AK147491.1 | - | 9 | 1736 | |||
| AK147878.1 | - | 9 | 1699 | |||
| AK154529.1 | - | 9 | 1716 | |||
| BC049900.1 | - | 3 | 425 | |||
| BC057351.1 | - | 9 | 1673 | |||
| BC060658.1 | - | 8 | 1304 | 30355406 | ||
| BC079590.1 | - | 9 | 1693 | |||

Arel1
Genomic Map