siRNA Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289572.1 | NP_001276501.1 | 6 | 935 | ||
| NM_001289574.1 | NP_001276503.1 | 7 | 1097 | |||
| NM_178914.4 | NP_849245.2 | 7 | 1031 | |||
| GenBank mRNA | AK036359.1 | - | 7 | 1015 | ||
| AK077500.1 | - | 7 | 942 | |||
| AK161308.1 | - | 6 | 881 | |||
| BC046960.1 | - | 7 | 989 | 6391450 | ||
| DQ196101.1 | - | 6 | 761 | |||

Spata7
Genomic Map