siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181072.3 | NP_851417.2 | 16, 17 | 2043 | ||
| GenBank mRNA | AK029145.1 | - | 16, 17 | 2045 | ||
| BC040786.1 | - | 17, 18 | 2099 | |||
| BC051391.1 | - | 16, 17 | 2025 | 5053090 | ||
| BY761514.1 | - | 3, 4 | 242 | |||

Myo1e
Genomic Map