siRNA Details
| Gene Symbol: Gp1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001999.1 | NP_001001999.1 | 1 | 1634 | ||
| NM_010327.2 | NP_034457.1 | 2 | 1396 | |||
| GenBank mRNA | AA222119.1 | - | 1 | 85 | ||
| BC053108.1 | - | 1 | 1634 | |||
| BC138156.1 | - | 1 | 1588 | 8861175 | ||
| BC145160.1 | - | 1 | 1588 | 9053711 | ||

Gp1bb
Genomic Map