siRNA Details
Gene Symbol: Sprr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY158991.1 | - | 2 | 189 | ||
BC130233.1 | - | 2 | 173 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sprr2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY158991.1 | - | 2 | 189 | ||
BC130233.1 | - | 2 | 173 |