siRNA Details
| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001448.2 | NP_001439.2 | 9 | 3495 | ||
| GenBank mRNA | AF030186.1 | - | 9 | 3496 | ||
| AF088063.1 | - | 1 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPC4| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001448.2 | NP_001439.2 | 9 | 3495 | ||
| GenBank mRNA | AF030186.1 | - | 9 | 3496 | ||
| AF088063.1 | - | 1 | 289 | |||