siRNA Details
| Gene Symbol: Trim29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023655.2 | NP_076144.2 | 9 | 2683 | ||
| GenBank mRNA | AK004195.1 | - | 2 | 937 | ||
| AK013219.1 | - | 8 | 1734 | |||
| AK028448.1 | - | 9 | 1916 | |||
| BC006699.1 | - | 9 | 2414 | |||

Trim29
Genomic Map