siRNA Details
| Gene Symbol: Ly6g6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033478.2 | NP_258439.1 | 2 | 116 | ||
| GenBank mRNA | AJ315548.1 | - | 2 | 116 | ||
| AJ315549.1 | - | 1 | 213 | |||
| AK043036.1 | - | 2 | 332 | |||
| BC131988.1 | - | 2 | 152 | 40130265 | ||
| BC131990.1 | - | 2 | 152 | 40130267 | ||
| BM933266.1 | - | 2 | 304 | |||

Ly6g6d
Genomic Map