siRNA Details
| Gene Symbol: Krtap6-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130856.2 | NP_570926.1 | 1 | 302 | ||
| GenBank mRNA | AF345298.1 | - | 1 | 284 | ||
| AK003281.1 | - | 1 | 303 | |||
| BC059086.1 | - | 1 | 286 | |||
| BC132612.1 | - | 1 | 266 | 40130889 | ||

Krtap6-5
Genomic Map