siRNA Details
| Gene Symbol: Erdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133362.2 | NP_579940.1 | 487 | |||
| GenBank mRNA | AJ007909.1 | - | 413 | |||
| AJ539223.1 | - | 425 | ||||
| AK051407.1 | - | 1016 | ||||
| BC018296.1 | - | 492 | 3499536 | |||
| BC058113.1 | - | 1 | 333 | 6820436 | ||
| BC080795.1 | - | 342 | 5006491 | |||
| BC082282.1 | - | 439 | ||||
| BC086616.1 | - | 3128 | ||||

Erdr1
Genomic Map