siRNA Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301347.1 | NP_001288276.1 | 5 | 725 | ||
| NM_001460.4 | NP_001451.2 | 7 | 1006 | |||
| GenBank mRNA | AK304385.1 | - | 5 | 633 | ||
| AK312877.1 | - | 7 | 912 | |||
| AL833218.1 | - | 7 | 895 | |||
| BC005894.1 | - | 7 | 912 | 4293031 | ||
| BT006979.1 | - | 6 | 848 | |||
| Y09267.1 | - | 7 | 908 | |||

FMO2
Genomic Map