siRNA Details
| Gene Symbol: Sprr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009265.3 | NP_033291.1 | 2 | 183 | ||
| GenBank mRNA | AK003398.1 | - | 2 | 184 | ||
| AK009752.1 | - | 2 | 184 | |||
| BC119288.1 | - | 2 | 176 | 8734037 | ||
| BC119290.1 | - | 2 | 176 | 8734039 | ||
| X91825.1 | - | 2 | 167 | |||

Sprr1b
Genomic Map