siRNA Details
| Gene Symbol: Defb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010030.1 | NP_034160.1 | 2 | 214 | ||
| GenBank mRNA | AJ011800.1 | - | 2 | 214 | ||
| BC109129.1 | - | 2 | 202 | 40038941 | ||
| BC109130.1 | - | 2 | 202 | 40038942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb2| Gene Symbol: Defb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010030.1 | NP_034160.1 | 2 | 214 | ||
| GenBank mRNA | AJ011800.1 | - | 2 | 214 | ||
| BC109129.1 | - | 2 | 202 | 40038941 | ||
| BC109130.1 | - | 2 | 202 | 40038942 | ||