siRNA Details
| Gene Symbol: Krtap5-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015808.1 | NP_056623.1 | 1 | 158 | ||
| GenBank mRNA | BC104355.1 | - | 158 | |||
| Gene Symbol: Gm4559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199309.1 | NP_001186238.1 | 1 | 95 | ||
| GenBank mRNA | BC104354.1 | - | 1 | 95 | ||
| BC120878.1 | - | 1 | 94 | |||

Krtap5-1
Genomic Map