siRNA Details
| Gene Symbol: Lce1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026822.1 | NP_081098.1 | 2 | 519 | ||
| GenBank mRNA | AK003951.1 | - | 2 | 519 | ||
| BC069979.1 | - | 2 | 496 | |||
| BC082291.1 | - | 2 | 491 | |||
| BC132596.1 | - | 1 | 470 | 40130873 | ||
| BC132598.1 | - | 1 | 470 | 40130875 | ||

Lce1b
Genomic Map