siRNA Details
Gene Symbol: Krtap5-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027844.4 | NP_082120.3 | 1 | 1164 | ||
GenBank mRNA | AK014785.1 | - | 2 | 631 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap5-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027844.4 | NP_082120.3 | 1 | 1164 | ||
GenBank mRNA | AK014785.1 | - | 2 | 631 |