siRNA Details
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 11 | 1764 | ||
NM_001286208.1 | NP_001273137.1 | 10 | 1709 | |||
NM_001286209.1 | NP_001273138.1 | 11 | 1656 | |||
NM_001481.2 | NP_001472.1 | 11 | 1641 | |||
XM_005256304.4 | XP_005256361.1 | 12 | 2296 | |||
XM_005256309.4 | XP_005256366.1 | 13 | 2213 | |||
XM_006721175.3 | XP_006721238.1 | 11 | 1694 | |||
XM_011522990.2 | XP_011521292.1 | 11 | 1715 | |||
XM_011522991.1 | XP_011521293.1 | 12 | 1870 | |||
XM_011522992.2 | XP_011521294.1 | 10 | 1663 | |||
XM_017023122.1 | XP_016878611.1 | 13 | 2021 | |||
XM_017023123.1 | XP_016878612.1 | 11 | 1720 | |||
XM_017023124.1 | XP_016878613.1 | 10 | 1507 | |||
XM_017023125.1 | XP_016878614.1 | 10 | 1639 | |||
GenBank mRNA | AF050079.1 | - | 11 | 1641 | ||
AK127378.1 | - | 2593 | ||||
AK293958.1 | - | 11 | 1671 | |||
AK297857.1 | - | 11 | 1624 | |||
BC104785.1 | - | 11 | 1626 | 8143788 | ||
BC112121.1 | - | 11 | 1626 | 8327589 | ||
BX647947.1 | - | 10 | 1628 | |||
CR749499.1 | - | 9 | 2320 |
